Learn More
Selleck Chemical LLC Gadobutrol S3497-5mg
SDP
Supplier:  Selleck Chemical LLC S34975MG
Encompass_Preferred
Gadobutrol (Gadovist Gadavist ZK 135079) is a non-ionic macrocyclic gadolinium-based contrast agent (GBCA) that is usedfor tissue contrast enhancement in magnetic resonance imaging (MRI)
Catalog No. 50-311-0809