Learn More
Apexbio Technology LLC Gadodiamide 122795-43-1 10mM (in 1mL H2O)
SDP
Supplier:  Apexbio Technology LLC B17655.1
Encompass_Preferred
Gadodiamide is a gadolinium-based paramagnetic agent commonly utilized in magnetic resonance imaging (MRI) studies As a nonionic hydrophilic chelate complex gadodiamide contains a central gadolinium (Gd) ion coordinated to an organic ligand to ensure solubility and stable circulation Upon intravenous administration its paramagnetic properties interact with protons in surrounding tissues shortening proton relaxation times (T1) thereby enhancing image contrast during MRI analysis This contrast enhancement enables detailed visualization of anatomical structures such as blood vessels tissues and pathological lesions within various biological systems Gadodiamide is widely employed in biomedical research to assess blood-brain barrier integrity evaluate vascular function and facilitate imaging studies focused on tissue perfusion and vascular abnormalities
Catalog No. 50-000-62009