Learn More
Strem, An Ascensus Company CAS# 14284-87-8. 5g. Gadolinium(III) acetylacetonate hydrate (99.9%-Gd) (REO). MFCD00150166
SDP
Supplier:  Strem, An Ascensus Company 9364015G
Encompass
CAS# 14284-87-8. 5g. Gadolinium(III) acetylacetonate hydrate (99.9%-Gd) (REO). MFCD00150166. Molecular Weight: 454.58. Molecular Formula: Gd(CH3COCHCOCH3)3∙XH2O. Color/form: off-white pwdr. Strem# 93-6401. http://www.strem.com/catalog/v/93-6401/
Catalog No. 50-901-16052