Learn More
Strem, An Ascensus Company CAS# 10138-52-0. 10g. Gadolinium(III) chloride, anhydrous (99.9%-Gd) (REO). MFCD00011024
SDP
Supplier:  Strem, An Ascensus Company 93641610G
Encompass
CAS# 10138-52-0. 10g. Gadolinium(III) chloride, anhydrous (99.9%-Gd) (REO). MFCD00011024. Molecular Weight: 263.61. Molecular Formula: GdCl3. Color/form: white pwdr. Strem# 93-6416. http://www.strem.com/catalog/v/93-6416/
Catalog No. 50-901-16058