Learn More
Cayman Chemical GadoterIdol 100mg
SDP
Supplier:  Cayman Chemical 33577100 MG
Encompass_Preferred
An MRI contrast agent; improves visualization of intraparenchymal and meningeal tumors compared to surrounding normal rat brain tissue in an orthotopic model of brain cancer at 0.25 mmol/kg; has been used to label MSCs in vitro to track their migration to spinal cord injury lesions in vivo
Catalog No. 50-294-7029