Learn More
TARGETMOL CHEMICALS INC Gadoteridol 100MG
SDP
Supplier:  TARGETMOL CHEMICALS INC T19333100MG
Encompass_Preferred
Also available in 1 mL, 10 mg, 25 mg, 50 mg, 200 mg and bulk. Please contact Fisher for quotes. Gadoteridol (Gd-HP-DO3A) is a gadolinium-based MRI contrast agent that can be used for central nervous system imaging. Purity 98.63%
Catalog No. 50-287-8410