Learn More
Medchemexpress LLC HY-16219 5mg Medchemexpress, Gadoxetate (Disodium) CAS:135326-22-6 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY162195MG
Encompass_Preferred
Medchemexpress, HY-16219 5mg Gadoxetate (Disodium) CAS:135326-22-6 Gadoxetate Disodium (Gd-EOB-DTPA Disodium) is a contrast agent in magnetic resonance imaging (MRI) of the hepatobiliary system, which accumulates in normal, functioning hepatocytes. Gadoxetate Disodium (Gd-EOB-DTPA Disodium) is used to evaluate focal liver lesions, such as hepatocellular carcinoma or liver metastasis on T1-weighted imaging [1] [2] [3] . Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-2764