Learn More
MSC Tape Logic Gaffers Tape, 60 yd, Blue, 1 roll
Supplier:  MSC 39588843
Encompass
Gaffers tape is constructed from poly coated cloth with a rubber-based adhesive, ideal for securing wires to carpet, wood, and concrete. It is extensively used in the entertainment industry for tasks such as bundling wires, taping down lighting and props, and masking silk-screen frames. The matte finish helps resist glare, making it suitable for stage applications.
  • Highly conformable and versatile for quick setups
  • Strong, industrial-grade construction
  • Removes easily without leaving residue
  • Suitable for indoor and outdoor use
  • Non-reflective surface
  • Abrasion-resistant
  • Water-resistant
  • Has an 11 mil thickness
  • Offers 45 lb. tensile strength
Catalog No. 50-191-9271