Learn More
Grainger Gage Pin Set ZZ .251 -.5 250 pcs.
Supplier:  Grainger 141F62
Encompass_Preferred
The Grainger Approved Gage Pin Set is a set of precision ground steel pins used to calibrate measuring tools and instruments. The set contains 9 pins ranging from 0.100" to 1.000" in 0.100" increments, with a tolerance of +/- 0.0002". The pins have a knurled grip area and are made from hardened and tempered alloy steel. They come in a fitted plastic case for storage and portability. The gage pin set allows for accurate calibration and inspection of gages, micrometers, dial indicators, and other precision measurement tools.
Catalog No. 50-275-2283