Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC GAK inhibitor 49 hydrochloride | 2930378-91-7 | 99.5% | 406.86 g·mol⁻¹ | C20H23ClN2O5 | 100MG
SDP

Supplier:  Medchemexpress LLC HY124793A100MG

Encompass_Preferred

GAK inhibitor 49 hydrochloride is a potent, ATP-competitive, and highly selective cyclin G-associated kinase (GAK) inhibitor supplied for laboratory research use. The compound is provided as the hydrochloride salt in milligram-scale pack sizes and is intended for in vitro and cellular studies; it is not for human or clinical use.

  • Potent ATP-competitive GAK inhibitor with reported Ki ≈ 0.54 nM and cellular IC50 ≈ 56 nM.
  • High purity (≈99.5%) suitable for research applications.
  • Supplied as the hydrochloride salt in milligram pack sizes (5-500 mg).
  • Solid appearance: white to yellow; molecular weight ≈ 406.86 g·mol⁻¹.
  • Store sealed at 4°C; in solvent, store at -80°C up to 6 months or -20°C up to 1 month.

Catalog No. 50-002-39307


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.