Learn More
TARGETMOL CHEMICALS INC GAL-021 5MG
SDP
Supplier:  TARGETMOL CHEMICALS INC T35575MG
Encompass_Preferred
Also available in 1mg 2mg 10mg 25mg 50mg 100mg 500mg and bulk. Please contact Fisher for quotes. GAL-021 an intravenous BKCa-channel blocker. purity: 99%
Catalog No. 50-257-6913