Learn More
Cayman Chemical GalactarIc AcId 500g
SDP
Supplier:  Cayman Chemical 36206500 G
Encompass_Preferred
A monosaccharide; increases mRNA expression of Runx2 in C3H/10T1/2 mouse mesenchymal stem cells from 5-10 µM; increases ALP levels in, and mineralization of, C3H/10T1/2 cells when used in combination with gelatin as a coating on a PLA scaffold at 10-20 µM
Catalog No. 50-291-1786