Learn More
Medchemexpress LLC Galactaric acid | 526-99-8 | MFCD00004239 | 98.0% | 210.14 g/mol | C6H10O8 | 500 G
SDP
Supplier:  Medchemexpress LLC HYW014410500G
Encompass_Preferred
Mucic acid (Galactaric acid) is a dicarboxylic sugar acid used as a research reagent and a building block in polymer chemistry and biomaterials. It has been investigated for effects on bone-related gene expression and is provided as a high-purity solid suitable for analytical and synthesis applications.
  • Used as a building block for polymer synthesis.
  • Reported to stimulate Runx2 mRNA expression.
  • Applied in polymer chemistry and bone tissue engineering research.
  • Provided as a high-purity solid for analytical use (typical purity 98%).
  • Well characterized: CAS 526-99-8, formula C6H10O8, molar mass 210.14 g/mol.
Catalog No. 50-003-02983