Learn More
Medchemexpress LLC Galactaric acid (mucic acid) | 526-99-8 | MFCD00004239 | 99.4% | 210.14 g/mol | C6H10O8 | 100 G
SDP
Supplier:  Medchemexpress LLC HYW014410100G
Encompass_Preferred
Mucic acid (galactaric acid) is a dicarboxylic sugar acid used as a building block in polymer synthesis and as an analytical reagent. It is used in polymer and bone tissue engineering research and has been reported to stimulate Runx2 mRNA expression.
  • Dicarboxylic sugar acid (C6H10O8) with molar mass 210.14 g/mol.
  • High purity suitable for analytical use (99.36%).
  • Useful building block for polymer synthesis and materials research.
  • Reported to stimulate Runx2 mRNA expression, relevant to bone tissue engineering studies.
  • Supplied as an analytical standard for research and testing.
Catalog No. 50-003-02911