Learn More
Medchemexpress LLC Galactose 1-phosphate Potassium salt | 19046-60-7 | 99.0% | 336.32 | 25 MG
SDP
Supplier:  Medchemexpress LLC 25MGHY113143A
Encompass_Preferred
Galactose 1-phosphate Potassium salt is an intermediate involved in galactose metabolism and nucleotide sugars. It acts as an inhibitor for several enzymatic activities, including phosphoglucomutase, glucose-6-phosphate phosphatase, glucose-6-phosphate dehydrogenase, glycogen phosphorylase, UDP-glucose pyrophosphorylase, and inositol monophosphatase. It is intended for research use only.
  • Intermediate in galactose metabolism and nucleotide sugars.
  • Inhibitor of various enzymatic activities (phosphoglucomutase, glucose-6-phosphate phosphatase, glucose-6-phosphate dehydrogenase, glycogen phosphorylase, UDP-glucose pyrophosphorylase, and inositol monophosphatase).
  • Available in solid and solution forms.
  • High purity (99.0%).
Catalog No. 50-004-39615