Learn More
Selleck Chemical LLC Galangin 3-methyl ether S3307-1mg
SDP
Supplier:  Selleck Chemical LLC S33071MG
Encompass_Preferred
Galangin 3-methyl ether (3-O-Methylgalangin 3-Methylgalangin) isolated from Lychnophora markgravii aerial parts shows strong antibacterial and antifungal effects
Catalog No. 50-310-8895