Learn More
Medchemexpress LLC Galanin Human HEK2 20ug
SDP
Supplier:  Medchemexpress LLC HYP7413220UG
Encompass_Preferred
Galanin exists as an endocrine hormone in the central and peripheral nervous systems and exerts its effects by binding to and activating G protein-coupled receptors (i e GALR1 GALR2 and GALR3) Galanin Protein Human (HEK293 Fc) is the recombinant human-derived Galanin protein expressed by HEK293 with C-hFc labeled tag
Catalog No. 50-001-79950