Learn More
Medchemexpress LLC Galanthamine-d6 | 1128109-00-1 | 99.5% | C17H15D6NO3 | 1 MG
SDP
Supplier:  Medchemexpress LLC HY76299S1MG
Encompass_Preferred
Galanthamine-d6 (Galantamine-d6) is a deuterium-labeled Galanthamine, known as a potent acetylcholinesterase (AChE) inhibitor with an IC50 of 500 nM. This compound is intended for research use only and can be utilized as a tracer or an internal standard for quantitative analysis by methods like NMR, GC-MS, or LC-MS. Stable heavy isotopes have been incorporated into drug molecules for quantitation during drug development, with deuteration potentially affecting pharmacokinetic and metabolic profiles.
  • Used as a tracer
  • Used as an internal standard for quantitative analysis
  • Deuterium-labeled compound
  • Potent acetylcholinesterase (AChE) inhibitor
Catalog No. 50-004-34529