Learn More
Enzo Life Sciences Galanthamine . hydrobromide (50mg). CAS: 1953-04-4
SDP
Supplier:  Enzo Life Sciences ALX550336M050
Encompass_Preferred
Specific, competitive and reversible acetylcholinesterase inhibitor. Alzheimer's disease therapeutic. Alternative name: Galantamine, Nivalin. Purity: ≥98% (HPLC). Solubility: Soluble in DMSO (10mg/ml) or 100% ethanol (4mg/ml); sparingly soluble in water. Long Term Storage: -20°C.
Catalog No. 50-200-9538