Learn More
Carbosynth LLC. Galardin TM, 5mg, MW: 388.47, Molecular formula: C20H28N4O4, CAS# 142880-36-2
SDP
Supplier:  Carbosynth LLC. INH3927PI5MG
Encompass_Preferred
Synonyms: N-[(2R)-2-(Hydroxamidocarbonylmethyl)-4-Methylpentanoyl]-L-Tryptophan Methylamide; GM6001; Ilomastat, Application: Inhibitor for Collagenases and MMPs, Storage Temperature: -20°C, References: D. Grobelny, I. Poncz, and R.E. Galardy, Biochemistry, 31, 7152 (1992)., R.J. Kok, et al., Bioconjug. Chem., 13, 128 (2002).
Catalog No. 50-168-5885