Learn More
Medchemexpress LLC Galaxolide 25g
SDP
Supplier:  Medchemexpress LLC 25GHYW100026
Encompass_Preferred
Galaxolide can induce estrogenic activity (Estrogen Receptor/ERR) trigger oxidative stress and genotoxicity and induce EROD and GST (Glutathione S-transferase) enzyme activity[1][2]
Catalog No. 50-002-06376