Learn More
ABclonal Technology Galectin 1/LGALS1 Rabbit mAb
SDP
Supplier:  ABclonal Technology A2121820UL
Encompass_Preferred
The galectins are a family of beta-galactoside-binding proteins implicated in modulating cell-cell and cell-matrix interactions This gene product may act as an autocrine negative growth factor that regulates cell proliferation
Catalog No. 50-000-06780