Learn More
Medchemexpress LLC Galectin-3/LGALS3 M 100ug | 100ug
SDP
Supplier:  Medchemexpress LLC HYP77684100UG
Encompass_Preferred
Galectin-3/LGALS3 protein binds IgE and cooperates with integrins -3 and -1 to promote endothelial cell migration It contributes to epithelial cell differentiation and acts as a splicing factor in the nucleus Galectin-3/LGALS3 Protein Mouse (HEK293 His) is the recombinant mouse-derived Galectin-3/LGALS3 protein expressed by HEK293 with C-His labeled tag
Catalog No. 50-001-87052