Learn More
Medchemexpress LLC Galectin-3/LGALS3 M 5ug | 5ug
SDP
Supplier:  Medchemexpress LLC HYP790795UG
Encompass_Preferred
Galectin-3 (LGALS3) is involved in multiple activities including IgE binding and signaling receptor binding It negatively regulates T-cell receptor signaling and modulates immune responses by modulating endocytosis and lymphocyte activation Galectin-3/LGALS3 Protein Mouse is the recombinant mouse-derived Galectin-3/LGALS3 protein expressed by E coli with tag free
Catalog No. 50-001-94544