Learn More
Cayman Chemical GalIc AcId-d2 50mg
SDP
Supplier:  Cayman Chemical 3513250 MG
Encompass_Preferred
An internal standard for the quantification of gallic acid by GC- or LC-MS
Catalog No. 50-291-1489