Learn More
Medchemexpress LLC Galiximab 10mg | 10mg
SDP
Supplier:  Medchemexpress LLC HYP9929310MG
Encompass_Preferred
Galiximab (IDEC-114) is a primatized monoclonal IgG1 antibody targeting CD80 Galiximab variable regions are primatized and the constant regions are humanized Galiximab induces antibody-dependent cellular cytotoxicity by blocking CD80-CD28 binding Galiximab has antitumor activity and can induce apoptosis in tumor cells Galiximab can be used in the research of relapsed Hodgkin lymphoma and solid tumors[1][2]
Catalog No. 50-003-23500