Learn More
Medchemexpress LLC GALK1-IN-1 | 669718-48-3 | 98.2% | 1 MG
SDP
Supplier:  Medchemexpress LLC HY1280471MG
Encompass_Preferred
GALK1-IN-1 (Compound 4) is a galactokinase inhibitor with an IC50 of 4.2129 μM. It is for research use only and not sold to patients.
Catalog No. 50-003-42894