Learn More
Apexbio Technology LLC Gallamine Triethiodide 65-29-2 100mg
SDP
Supplier:  Apexbio Technology LLC B1610100
Encompass_Preferred
Gallamine Triethiodide (CAS 65-29-2) is a synthetic neuromuscular blocking agent that acts as a nondepolarizing antagonist at nicotinic acetylcholine receptors located at the neuromuscular junction By competitively inhibiting acetylcholine binding it prevents depolarization of muscle fibers thereby inducing skeletal muscle relaxation Gallamine Triethiodide is utilized in research to investigate neuromuscular transmission receptor pharmacology and the effects of neuromuscular blockade on physiological processes Its application extends to studies on the modulation of synaptic function and the evaluation of novel muscle relaxant compounds
Catalog No. 50-000-91278