Learn More
Strem, An Ascensus Company CAS# 14405-43-7. 5g. Gallium(III) acetylacetonate (99.99+%-Ga) PURATREM. MFCD00013492
SDP
Supplier:  Strem, An Ascensus Company 9331305G
Encompass
CAS# 14405-43-7. 5g. Gallium(III) acetylacetonate (99.99+%-Ga) PURATREM. MFCD00013492. Molecular Weight: 367.05. Molecular Formula: Ga(CH3COCHCOCH3)3. Color/form: white to pale yellow pwdr. Strem# 93-3130. http://www.strem.com/catalog/v/93-3130/
Catalog No. 50-901-15034