Learn More
Strem, An Ascensus Company CAS# 13450-90-3. 5g. Gallium(III) chloride, anhydrous, granular (99.999%-Ga) PURATREM. MFCD00011018
SDP
Supplier:  Strem, An Ascensus Company 9331405G
Encompass
CAS# 13450-90-3. 5g. Gallium(III) chloride, anhydrous, granular (99.999%-Ga) PURATREM. MFCD00011018. Molecular Weight: 176.03. Molecular Formula: GaCl3. Color/form: white crystalline pwdr. Strem# 93-3140. http://www.strem.com/catalog/v/93-3140/
Catalog No. NC0457436