Learn More
Strem, An Ascensus Company CAS# 13450-91-4. 5g. Gallium(III) iodide, anhydrous, 99%. MFCD00016107
SDP
Supplier:  Strem, An Ascensus Company 9331105G
Encompass
CAS# 13450-91-4. 5g. Gallium(III) iodide, anhydrous, 99%. MFCD00016107. Molecular Weight: 450.44. Molecular Formula: GaI3. Color/form: yellow pwdr. Strem# 93-3110. http://www.strem.com/catalog/v/93-3110/
Catalog No. NC1434358