Learn More
Strem, An Ascensus Company CAS# 69365-72-6. 5g. Gallium(III) nitrate hydrate (99.99%-Ga) PURATREM. MFCD00149723
SDP
Supplier:  Strem, An Ascensus Company 9331055G
Encompass
CAS# 69365-72-6. 5g. Gallium(III) nitrate hydrate (99.99%-Ga) PURATREM. MFCD00149723. Molecular Weight: 255.73. Molecular Formula: Ga(NO3)3∙XH2O. Color/form: white xtl. Strem# 93-3105. http://www.strem.com/catalog/v/93-3105/
Catalog No. 50-901-15021