Learn More
Medchemexpress LLC (+)-gallocatechin | 970-73-0 | 99.7% | 10 MG

Supplier: Medchemexpress LLC HYN0521A10MG
(+)-Gallocatechin is a polyphenol isolated from green tea with reported anticancer activity. Supplied as a high-purity enantiomer for laboratory research, it is suitable for in vitro cell-based assays and biochemical studies.
- High purity (≈99.7%) suitable for sensitive assays.
- Molecular formula C15H14O7 and molecular weight 306.27 g/mol.
- Soluble in DMSO (100 mg/mL) and partially soluble in water (3.33 mg/mL); ultrasonic assistance may be required.
- Reported to inhibit cell proliferation and induce apoptosis in certain cancer cell lines.
- Store at 4°C and protect from light; in solvent: -80°C (6 months) or -20°C (1 month).
- For research use only; not for human or clinical applications.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.