Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Gallocyanine (chloride) | 1562-85-2 | 92.0% | 50 MG
SDP

Supplier:  Medchemexpress LLC HYD096150MG

Encompass_Preferred

Gallocyanine chloride is a synthetic blue dyestuff that can be used as a potential agent for the research of Alzheimer's disease and related neurodegenerative tauopathies. It inhibits DKK1/LRP6 interaction (IC50=6.38 μM), activates the Wnt signaling pathway, and causes β-catenin accumulation. This compound also exhibits anti-metastasis, anti-inflammatory, and anti-fibrosis activities. It can further be used as a fluorescent probe for the detection of superoxide anion radicals.

  • Molecular weight: 336.73
  • Formula: C15H13ClN2O5
  • Appearance: Solid
  • Color: Brown to black
  • Shipping: Room temperature in continental US; may vary elsewhere
  • Storage: 4°C, sealed storage, away from moisture and light
  • In solvent storage: -80°C, 6 months; -20°C, 1 month (sealed storage, away from moisture and light)

Catalog No. 50-004-11051


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.