Learn More
TARGETMOL CHEMICALS INC GALLOFLAVIN 10MG
SDP
Supplier:  TARGETMOL CHEMICALS INC T2233610MG
Encompass_Preferred
Also available in 1 mg 5 mg 25 mg 50 mg 100 mg and bulk. Please contact Fisher for quotes. Galloflavin inhibits both the A and B isoforms of lactate dehydrogenase with Ki of 5.46 uM and 15.06 uM respectively. Galloflavin is also a potential anticancer agent. purity: 96%
Catalog No. 50-257-7191