Learn More
Medchemexpress LLC Galloflavin | 568-80-9 | MFCD26941292 | >=95.0% | 278.17 g·mol⁻¹ | C12H6O8 | 5 MG
SDP
Supplier:  Medchemexpress LLC HYW0401185MG
Encompass_Preferred
Galloflavin is a small-molecule research compound that potently inhibits lactate dehydrogenase (LDH), blocking glycolysis and lowering cellular ATP levels. It inhibits both LDH-A and LDH-B isoforms and is used in cellular and in vivo studies to investigate metabolic pathways and cancer cell metabolism.
  • Inhibits lactate dehydrogenase A and B isoforms.
  • Blocks glycolysis and decreases ATP production in cells.
  • Demonstrates antiproliferative effects in cancer cell models.
  • Available as a powder and as a DMSO solution for in vitro use.
  • Soluble in DMSO at 5 mg/mL; multiple in vivo formulation options described.
  • Stable when stored as powder at -20°C for long-term storage.
Catalog No. 50-003-01219