Learn More
Cayman Chemical Galloflavin, 568-80-9, 1 mg
SDP
Supplier:  Cayman Chemical 148461
Encompass_Preferred
An LDH inhibitor (Kis = 5.46 and 15.06 µM, for the A and B isoforms, respectively) that prevents proliferation of many different cancer cells by blocking glycolysis and ATP production
Catalog No. NC1176296