Learn More
Medchemexpress LLC Gallopamil hydrochloride | 16662-46-7 | 99.15% | C28H41ClN2O5 | 1 ML
SDP
Supplier:  Medchemexpress LLC HY14276A10MM/1ML
Encompass_Preferred
Gallopamil hydrochloride (Methoxyverapamil hydrochloride), a methoxy derivative of Verapamil, is a phenylalkylamine calcium antagonist. It inhibits acid secretion in a concentration-dependent manner with an IC50 of 10.9 μM. It is also a potent antiarrhythmic and vasodilator agent.
  • Also known as: Methoxyverapamil hydrochloride
  • Methoxy derivative of Verapamil
  • Phenylalkylamine calcium antagonist
  • Inhibits acid secretion in a concentration-dependent manner with an IC50 of 10.9 μM
  • Potent antiarrhythmic and vasodilator agent
Catalog No. 50-003-53584