Learn More
Medchemexpress LLC GalNAc-L96 | 1159408-62-4 | 97.0% | 2608.95 | 50 MG
SDP
Supplier:  Medchemexpress LLC HY14701250MG
Encompass_Preferred
GalNAc-L96 is a triantennary N-acetylgalactosamine that functions as a ligand for the asialoglycoprotein receptor (ASGPR). It can be utilized in the synthesis of modified siRNA and is applicable for oligonucleotide delivery.
  • It is a triantennary N-acetylgalactosamine.
  • Functions as a ligand for the asialoglycoprotein receptor.
  • Can be used to synthesize modified siRNA.
  • Applicable for oligonucleotide delivery.
  • Binds to asialoglycoprotein receptors with high affinity.
Catalog No. 50-003-59887