Learn More
TARGETMOL CHEMICALS INC GALOCITABINE 5MG
SDP
Supplier:  TARGETMOL CHEMICALS INC T273995MG
Encompass_Preferred
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg and bulk. Please contact Fisher for quotes. Galocitabine (Neofrutulon) a thymidylate synthase inhibitor is used potentially for the treatment of cancer. purity: 99%
Catalog No. 50-258-0618