Learn More
TARGETMOL CHEMICALS INC GALUNISERTIB 50MG
SDP
Supplier:  TARGETMOL CHEMICALS INC T251050MG
Encompass_Preferred
Also available in 5 mg 10 mg 25 mg 100 mg 200 mg 500 mg 1000 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Galunisertib (LY2157299) is an orally available inhibitor of TGF(beta)RI kinase (IC50 56 nM) with potential antineoplastic activity. purity: 99%
Catalog No. 50-257-7103