Learn More
Medchemexpress LLC Gambogenic acid | 173932-75-7 | 50 MG
SDP
Supplier:  Medchemexpress LLC HYN502450MG
Encompass_Preferred
Gambogenic acid is an active ingredient isolated from gamboge. It acts as an effective inhibitor of EZH2 by specifically and covalently binding to Cys668 within the EZH2-SET domain, which induces EZH2 ubiquitination. Gambogenic acid is used for cancer research.
  • Active ingredient isolated from gamboge
  • Effective inhibitor of EZH2
  • Specifically and covalently binds to Cys668 within EZH2-SET domain
  • Induces EZH2 ubiquitination
  • Used for cancer research
Catalog No. 50-004-18065