Learn More
Selleck Chemical LLC Gambogic Acid S2448-1g
SDP
Supplier:  Selleck Chemical LLC S24481G
Encompass_Preferred
Gambogic Acid (Guttatic Acid Guttic Acid Beta-Guttiferrin) activates caspases with EC50 of 0 78-1 64 M and competitively inhibits Bcl-XL Bcl-2 Bcl-W Bcl-B Bfl-1 and Mcl-1 with IC50 of 1 47 1 21 2 02 0 66 1 06 and 0 79 M respectively
Catalog No. 50-311-2925