Learn More
Enzo Life Sciences Gambogic acid (25mg). CAS: 2752-65-0
SDP
Supplier:  Enzo Life Sciences BMLAP3050025
Encompass_Preferred
Gambogic Acid is a natural product isolated from Garcinia Hurburyi. It induces apoptosis in several tumor cell lines including T47D breast cancer cells and MGC-803 human gastric carcinoma cells but not in normal cells. Apoptosis induction proceeds via upregulation of bax and inhibition of bcl-2 expression.The target of gambogic acid has been identified as the transferrin receptor. Purity: ≥95% (TLC). Solubility: Soluble in DMSO (25 mg/ml) or ethanol (25 mg/ml). Long Term Storage: -20°C.
Catalog No. 50-201-0315