Learn More
Alkali Scientific Gamborg's B5 Medium Vitamin Mixture, 11.2 Grams of Powder, Makes 100 Milliliters of Solution
SDP
Supplier:  Alkali Scientific MBO001CP
Encompass
Gamborg's B5 Medium Vitamin Mixture is a high-quality powder formulation used to prepare 100 milliliters of plant tissue culture media. Specifically designed for plant cell growth, it contains essential vitamins and nutrients necessary for successful cultivation of plant cells in vitro. The mixture is commonly used in plant biotechnology for culturing various plant species, including those used in agricultural research and genetic studies. The powder is highly soluble in water, providing an ideal nutrient solution for optimal plant cell growth. This medium is an essential tool in laboratories studying plant development, regeneration, and transformation processes.
Catalog No. 50-006-60120