Learn More
Medchemexpress LLC Gamitrinib TPP hexafluorophosphate | 1131626-47-5 | MFCD31715433 | 99.7% | 1036.03 g/mol | C52H65F6N3O8P2 | 1 ML
SDP
Supplier:  Medchemexpress LLC HY102007A10MM 1ML
Encompass_Preferred
Gamitrinib TPP hexafluorophosphate is a mitochondria-targeted HSP90 inhibitor that localizes to the mitochondrial matrix and exhibits anticancer activity in preclinical studies. It is supplied as the hexafluorophosphate salt and is available in solution and solid formats for research use.
  • Targets mitochondrial HSP90 and disrupts mitochondrial proteostasis.
  • Localizes to the mitochondrial matrix via triphenylphosphonium conjugation.
  • Provided as a hexafluorophosphate salt for chemical stability.
  • Available as a 10 mM solution in DMSO or in multiple solid quantities for experimental flexibility.
  • High purity suitable for biochemical and cellular assays.
  • Molecular weight 1036.03 g/mol; molecular formula C52H65F6N3O8P2.
Catalog No. 50-002-47434