Learn More
Abcam gamma-DGG (gamma-D-Glutamylglycine) (mM/ml), Ionotropic glutamate antagonist, 100MM_ML
Supplier:  Abcam AB146702100MM_ML
Encompass_Preferred
MW 204.18 Da, Purity >99%. Ionotropic glutamate receptor antagonist that has been used as a rapidly dissociating, low affinity competitive antagonist at AMPA receptors.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-270-4517