Learn More
Medchemexpress LLC Gamma-glutamylcysteine trifluoroacetate | 283159-88-6 | MFCD00133357 | >95.0% | 364.30 g/mol | C10H15F3N2O7S | 5 MG

Supplier: Medchemexpress LLC HY113402A5MG
Gamma-glutamylcysteine trifluoroacetate (TFA) is the TFA salt of the dipeptide gamma-glutamylcysteine, a biochemical precursor to glutathione (GSH). Supplied as a white to off-white solid for research and analytical applications, it is used to study glutathione metabolism, antioxidant pathways, and related biochemical assays.
- TFA salt of gamma-glutamylcysteine, a glutathione precursor.
- Purity >95%.
- Chemical formula C10H15F3N2O7S; molecular weight 364.30 g/mol.
- Supplied as a solid suitable for solution preparation in DMSO or aqueous media.
- Intended for research and analytical use only.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.