Learn More
Medchemexpress LLC Gamma-glutamylglutamine | 10148-81-9 | MFCD00038475 | 98.5% | 275.26 g/mol | C10H17N3O6 | 5 MG

Supplier: Medchemexpress LLC HY1130035MG
H-γ-Glu-Gln-OH (γ-glutamylglutamine) is a small, gamma-linked dipeptide used as a biochemical research reagent. It is a hydrophilic compound supplied as a white to off-white solid and commonly used in peptide chemistry, metabolic studies, and analytical assays.
- Molecular formula: C10H17N3O6.
- Molecular weight: 275.26 g/mol.
- Appearance: white to off-white solid.
- Purity: 98.5%.
- Package size: 5 mg.
- Typical applications: peptide synthesis support, metabolic and biochemical research, analytical reference material.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.