Learn More
Medchemexpress LLC Gamma-glutamylphenylalanine | 7432-24-8 | 99.8% | 294.30 g/mol | C14H18N2O5 | 10 MG

Supplier: Medchemexpress LLC HY101399R10MG
γ-Glu-Phe (γ-glutamylphenylalanine) is an analytical reference standard supplied as a small-quantity solid for laboratory and analytical use. It is used for endogenous metabolite quantification, method development, and quality control. The certificate of analysis reports molecular formula C14H18N2O5, molecular weight 294.30 g/mol, and HPLC purity of 99.81%. Storage is recommended at 4°C, sealed and protected from light and moisture.
- Suitable for analytical reference and method validation.
- Reported HPLC purity of 99.8%.
- Molecular formula C14H18N2O5 and molecular weight 294.30 g/mol.
- Supplied as a sealed solid in a brown glass vial for stability.
- Storage at 4°C, protected from light and moisture.
- Intended for research and analytical laboratory use.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.